Assay Details
| Gene Symbol: mms-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072056.5 | NP_504457.2 | 7 - 8 | 1022 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mms-19| Gene Symbol: mms-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072056.5 | NP_504457.2 | 7 - 8 | 1022 | 73 | |
Set Membership: |
|