Assay Details
| Gene Symbol: syp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072061.7 | NP_504462.1 | 2 - 3 | 502 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

syp-2| Gene Symbol: syp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072061.7 | NP_504462.1 | 2 - 3 | 502 | 97 | |
Set Membership: |
|