Assay Details
| Gene Symbol: klc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028492.4 | NP_001023663.1 | 5 - 6 | 1588 | 98 | |
| NM_001028494.5 | NP_001023665.1 | 4 - 5 | 1494 | 98 | ||
More Information
Set Membership: |
|

klc-2
Genomic Map