Assay Details
| Gene Symbol: cyn-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129598.3 | NP_001123070.1 | 3 - 4 | 576 | 97 | |
| NM_001129599.4 | NP_001123071.1 | 3 - 4 | 606 | 97 | ||
More Information
Set Membership: |
|

cyn-17
Genomic Map