Assay Details
| Gene Symbol: fut-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072154.8 | NP_504555.2 | 1 - 2 | 113 | 94 | |
| GenBank mRNA | AJ512486.1 | - | 1 - 2 | 91 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fut-8| Gene Symbol: fut-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072154.8 | NP_504555.2 | 1 - 2 | 113 | 94 | |
| GenBank mRNA | AJ512486.1 | - | 1 - 2 | 91 | 94 | |
Set Membership: |
|