Assay Details
| Gene Symbol: fut-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072154.8 | NP_504555.2 | 2 - 3 | 238 | 118 | |
| GenBank mRNA | AJ512486.1 | - | 2 - 3 | 216 | 118 | |
More Information
Set Membership: |
|

fut-8
Genomic Map