Assay Details
| Gene Symbol: atf-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072175.5 | NP_504576.1 | 3 - 4 | 460 | 80 | |
| Gene Symbol: cwf-19L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072176.6 | NP_504577.1 | 3 - 4 | 363 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_072176.6 transcripts.
Set Membership: |
|

atf-8
Genomic Map