Assay Details
Gene Symbol: spe-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356661.4 | NP_001343858.1 | 9 | 1354 | 79 | |
NM_001356662.3 | NP_001343857.1 | 9 - 10 | 1411 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
