Assay Details
| Gene Symbol: atp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380669.2 | NP_001367908.1 | 8 - 9 | 1711 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

atp-4| Gene Symbol: atp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380669.2 | NP_001367908.1 | 8 - 9 | 1711 | 74 | |
Set Membership: |
|