Assay Details
| Gene Symbol: dpy-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383321.1 | 4 - 5 | 598 | 72 | ||
| GenBank mRNA | AF250045.1 | - | 3 - 4 | 445 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-11| Gene Symbol: dpy-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383321.1 | 4 - 5 | 598 | 72 | ||
| GenBank mRNA | AF250045.1 | - | 3 - 4 | 445 | 72 | |
Set Membership: |
|