Assay Details
| Gene Symbol: fut-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072270.2 | NP_504671.1 | 3 - 4 | 314 | 164 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fut-2| Gene Symbol: fut-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072270.2 | NP_504671.1 | 3 - 4 | 314 | 164 | |
Set Membership: |
|