Assay Details
| Gene Symbol: scd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072284.4 | NP_504685.3 | 3 - 4 | 449 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scd-2| Gene Symbol: scd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072284.4 | NP_504685.3 | 3 - 4 | 449 | 98 | |
Set Membership: |
|