Assay Details
| Gene Symbol: spn-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072316.6 | NP_504717.1 | 2 - 3 | 170 | 64 | |
| GenBank mRNA | AB052819.1 | - | 2 - 3 | 173 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

spn-4| Gene Symbol: spn-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072316.6 | NP_504717.1 | 2 - 3 | 170 | 64 | |
| GenBank mRNA | AB052819.1 | - | 2 - 3 | 173 | 64 | |
Set Membership: |
|