Assay Details
| Gene Symbol: lgc-54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356670.2 | NP_001343648.1 | 6 - 7 | 1015 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lgc-54| Gene Symbol: lgc-54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356670.2 | NP_001343648.1 | 6 - 7 | 1015 | 93 | |
Set Membership: |
|