Assay Details
| Gene Symbol: lipl-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072354.4 | NP_504755.3 | 3 - 4 | 765 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lipl-6| Gene Symbol: lipl-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072354.4 | NP_504755.3 | 3 - 4 | 765 | 67 | |
Set Membership: |
|