Assay Details
| Gene Symbol: ptr-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072359.4 | NP_504760.1 | 9 - 10 | 1764 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ptr-15| Gene Symbol: ptr-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072359.4 | NP_504760.1 | 9 - 10 | 1764 | 110 | |
Set Membership: |
|