Assay Details
| Gene Symbol: mys-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072395.4 | NP_504796.1 | 3 - 4 | 666 | 124 | |
| GenBank mRNA | AY551963.1 | - | 3 - 4 | 687 | 124 | |
More Information
Set Membership: |
|

mys-1
Genomic Map