Assay Details
| Gene Symbol: C13A2.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072446.4 | NP_504847.1 | 1 - 2 | 355 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C13A2.6| Gene Symbol: C13A2.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072446.4 | NP_504847.1 | 1 - 2 | 355 | 82 | |
Set Membership: |
|