Assay Details
| Gene Symbol: F07G11.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356693.2 | NP_001343621.1 | 2 - 3 | 365 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F07G11.3| Gene Symbol: F07G11.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356693.2 | NP_001343621.1 | 2 - 3 | 365 | 95 | |
Set Membership: |
|