Assay Details
| Gene Symbol: ddo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383337.1 | 2 - 3 | 743 | 80 | ||
| GenBank mRNA | AB275892.1 | - | 1 - 2 | 183 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ddo-2| Gene Symbol: ddo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383337.1 | 2 - 3 | 743 | 80 | ||
| GenBank mRNA | AB275892.1 | - | 1 - 2 | 183 | 80 | |
Set Membership: |
|