Assay Details
| Gene Symbol: ddo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383337.1 | 3 - 4 | 948 | 67 | ||
| NR_136371.1 | 2 - 3 | 399 | 67 | |||
| GenBank mRNA | AB275892.1 | - | 2 - 3 | 388 | 67 | |
More Information
Set Membership: |
|

ddo-2
Genomic Map