Assay Details
| Gene Symbol: ddo-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383337.1 | 7 - 8 | 1462 | 66 | ||
| NR_136371.1 | 6 - 7 | 913 | 66 | |||
| GenBank mRNA | AB275892.1 | - | 6 - 7 | 902 | 66 | |
More Information
Set Membership: |
|

ddo-2
Genomic Map