Assay Details
| Gene Symbol: D1014.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072695.2 | NP_505096.1 | 4 - 5 | 699 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

D1014.6| Gene Symbol: D1014.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072695.2 | NP_505096.1 | 4 - 5 | 699 | 120 | |
Set Membership: |
|