Assay Details
| Gene Symbol: spr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072697.5 | NP_505098.1 | 3 - 4 | 499 | 135 | |
| GenBank mRNA | AF548624.1 | - | 3 - 4 | 499 | 135 | |
| AY160228.1 | - | 3 - 4 | 496 | 135 | ||
More Information
Set Membership: |
|

spr-1
Genomic Map