Assay Details
| Gene Symbol: spr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072697.5 | NP_505098.1 | 4 - 5 | 669 | 121 | |
| GenBank mRNA | AF548624.1 | - | 4 - 5 | 669 | 121 | |
| AY160228.1 | - | 4 - 5 | 666 | 121 | ||
More Information
Set Membership: |
|

spr-1
Genomic Map