Assay Details
| Gene Symbol: srp-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028652.7 | NP_001023823.1 | 3 - 4 | 571 | 121 | |
| NM_001028653.5 | NP_001023824.1 | 2 - 3 | 463 | 121 | ||
| NM_001129452.4 | NP_001122924.1 | 3 - 4 | 484 | 121 | ||
| NM_001383352.1 | 3 - 4 | 566 | 121 | |||
| GenBank mRNA | AY525083.1 | - | 2 - 3 | 463 | 121 | |
| AY525084.1 | - | 2 - 3 | 463 | 121 | ||
| AY525085.1 | - | 2 - 3 | 463 | 121 | ||
More Information
Set Membership: |
|

srp-7
Genomic Map