Assay Details
| Gene Symbol: air-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072718.10 | NP_505119.1 | 6 - 7 | 792 | 92 | |
| GenBank mRNA | AF071206.1 | - | 6 - 7 | 809 | 92 | |
More Information
Set Membership: |
|

air-1
Genomic Map