Assay Details
| Gene Symbol: gpa-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072756.5 | NP_505157.1 | 4 - 5 | 391 | 120 | |
| GenBank mRNA | AY008125.1 | - | 4 - 5 | 382 | 120 | |
More Information
Set Membership: |
|

gpa-2
Genomic Map