Assay Details
| Gene Symbol: gpa-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072756.5 | NP_505157.1 | 6 - 7 | 607 | 99 | |
| GenBank mRNA | AY008125.1 | - | 6 - 7 | 598 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gpa-2| Gene Symbol: gpa-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072756.5 | NP_505157.1 | 6 - 7 | 607 | 99 | |
| GenBank mRNA | AY008125.1 | - | 6 - 7 | 598 | 99 | |
Set Membership: |
|