Assay Details
| Gene Symbol: pxn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072787.4 | NP_505188.3 | 8 - 9 | 1153 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pxn-1| Gene Symbol: pxn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072787.4 | NP_505188.3 | 8 - 9 | 1153 | 116 | |
Set Membership: |
|