Assay Details
| Gene Symbol: F45F2.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072791.6 | NP_505192.1 | 6 - 7 | 2277 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F45F2.10| Gene Symbol: F45F2.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072791.6 | NP_505192.1 | 6 - 7 | 2277 | 91 | |
Set Membership: |
|