Assay Details
| Gene Symbol: rpm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072866.4 | NP_505267.1 | 14 - 15 | 11089 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpm-1| Gene Symbol: rpm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072866.4 | NP_505267.1 | 14 - 15 | 11089 | 61 | |
Set Membership: |
|