Assay Details
| Gene Symbol: gck-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313269.3 | NP_001300198.1 | 1 - 2 | 76 | 80 | |
| NM_001383362.1 | 1 - 2 | 88 | 80 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gck-1| Gene Symbol: gck-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313269.3 | NP_001300198.1 | 1 - 2 | 76 | 80 | |
| NM_001383362.1 | 1 - 2 | 88 | 80 | |||
Set Membership: |
|