Assay Details
| Gene Symbol: gck-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028970.3 | NP_001024141.1 | 4 - 5 | 751 | 77 | |
| NM_001313268.2 | NP_001300197.1 | 4 - 5 | 1020 | 77 | ||
| NM_001383362.1 | 4 - 5 | 635 | 77 | |||
| NM_072909.5 | NP_505310.2 | 4 - 5 | 1077 | 77 | ||
More Information
Set Membership: |
|

gck-1
Genomic Map