Assay Details
| Gene Symbol: gck-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028970.3 | NP_001024141.1 | 5 - 6 | 950 | 153 | |
| NM_001313268.2 | NP_001300197.1 | 5 - 6 | 1219 | 153 | ||
| NM_001313269.3 | NP_001300198.1 | 5 - 6 | 819 | 153 | ||
| NM_001313270.4 | NP_001300199.1 | 5 - 6 | 947 | 153 | ||
| NM_001313271.4 | NP_001300200.1 | 6 | 869 | 153 | ||
| NM_001383362.1 | 5 - 6 | 834 | 153 | |||
| NM_072909.5 | NP_505310.2 | 5 - 6 | 1276 | 153 | ||
| NM_072910.8 | NP_505311.3 | 5 - 6 | 1273 | 153 | ||
More Information
Set Membership: |
|

gck-1
Genomic Map