Assay Details
| Gene Symbol: gck-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028970.3 | NP_001024141.1 | 7 - 8 | 1248 | 63 | |
| NM_001313268.2 | NP_001300197.1 | 7 - 8 | 1517 | 63 | ||
| NM_001313269.3 | NP_001300198.1 | 7 | 1117 | 63 | ||
| NM_001313270.4 | NP_001300199.1 | 7 - 8 | 1245 | 63 | ||
| NM_001313271.4 | NP_001300200.1 | 8 | 1167 | 63 | ||
| NM_001383362.1 | 7 - 8 | 1132 | 63 | |||
| NM_072909.5 | NP_505310.2 | 7 - 8 | 1574 | 63 | ||
| NM_072910.8 | NP_505311.3 | 7 - 8 | 1571 | 63 | ||
More Information
Set Membership: |
|

gck-1
Genomic Map