Assay Details
| Gene Symbol: gck-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313269.3 | NP_001300198.1 | 3 - 4 | 472 | 117 | |
| NM_001313270.4 | NP_001300199.1 | 3 - 4 | 600 | 117 | ||
| NM_001313271.4 | NP_001300200.1 | 4 | 522 | 117 | ||
| NM_072910.8 | NP_505311.3 | 3 - 4 | 926 | 117 | ||
More Information
Set Membership: |
|

gck-1
Genomic Map