Assay Details
| Gene Symbol: swm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072945.3 | NP_505346.1 | 2 - 3 | 132 | 57 | |
| Gene Symbol: C25E10.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383365.1 | 1 - 2 | 52 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383365.1 transcripts.
Set Membership: |
|

swm-1
Genomic Map