Assay Details
Gene Symbol: F21C10.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072961.6 | NP_505362.2 | 2 - 3 | 289 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F21C10.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072961.6 | NP_505362.2 | 2 - 3 | 289 | 81 |
Set Membership: |
![]() ![]() ![]() ![]() |