Assay Details
| Gene Symbol: flp-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073043.5 | NP_505444.1 | 2 - 3 | 264 | 82 | |
| GenBank mRNA | AF042392.1 | - | 2 - 3 | 261 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

flp-6| Gene Symbol: flp-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073043.5 | NP_505444.1 | 2 - 3 | 264 | 82 | |
| GenBank mRNA | AF042392.1 | - | 2 - 3 | 261 | 82 | |
Set Membership: |
|