Assay Details
| Gene Symbol: flp-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073043.5 | NP_505444.1 | 3 - 4 | 344 | 122 | |
| GenBank mRNA | AF042392.1 | - | 3 - 4 | 341 | 122 | |
More Information
Set Membership: |
|

flp-6
Genomic Map