Assay Details
| Gene Symbol: mdf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073061.5 | NP_505462.1 | 2 - 3 | 770 | 108 | |
| GenBank mRNA | AF239998.1 | - | 2 - 3 | 768 | 108 | |
More Information
Set Membership: |
|

mdf-1
Genomic Map