Assay Details
| Gene Symbol: ehbp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073067.4 | NP_505468.1 | 7 - 8 | 1799 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ehbp-1| Gene Symbol: ehbp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073067.4 | NP_505468.1 | 7 - 8 | 1799 | 72 | |
Set Membership: |
|