Assay Details
| Gene Symbol: atl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269199.3 | NP_001256128.1 | 1 - 2 | 358 | 86 | |
| NM_073086.5 | NP_505487.3 | 13 - 14 | 4812 | 86 | ||
More Information
Set Membership: |
|

atl-1
Genomic Map