Assay Details
| Gene Symbol: atl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269199.3 | NP_001256128.1 | 3 - 4 | 1513 | 84 | |
| NM_073086.5 | NP_505487.3 | 15 - 16 | 5967 | 84 | ||
More Information
Set Membership: |
|

atl-1
Genomic Map