Assay Details
| Gene Symbol: atl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269199.3 | NP_001256128.1 | 4 - 5 | 2163 | 95 | |
| NM_073086.5 | NP_505487.3 | 16 - 17 | 6617 | 95 | ||
More Information
Set Membership: |
|

atl-1
Genomic Map