Assay Details
| Gene Symbol: srab-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028963.3 | NP_001024134.1 | 4 - 5 | 638 | 71 | |
| NR_131697.1 | 4 - 5 | 673 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srab-26| Gene Symbol: srab-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028963.3 | NP_001024134.1 | 4 - 5 | 638 | 71 | |
| NR_131697.1 | 4 - 5 | 673 | 71 | |||
Set Membership: |
|