Assay Details
| Gene Symbol: wdr-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348580.3 | NP_001335505.1 | 2 - 3 | 192 | 90 | |
| NM_001380736.2 | NP_001368633.1 | 1 - 2 | 75 | 90 | ||
| NM_001383378.1 | 2 - 3 | 190 | 90 | |||
More Information
Set Membership: |
|

wdr-20
Genomic Map