Assay Details
| Gene Symbol: wdr-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348580.3 | NP_001335505.1 | 11 | 2034 | 70 | |
| NM_001380736.2 | NP_001368633.1 | 9 - 10 | 1917 | 70 | ||
| NM_001383378.1 | 10 - 11 | 2032 | 70 | |||
More Information
Set Membership: |
|

wdr-20
Genomic Map