Assay Details
| Gene Symbol: akt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028474.7 | NP_001023645.1 | 5 - 6 | 816 | 92 | |
| GenBank mRNA | AF072379.1 | - | 5 - 6 | 816 | 92 | |
More Information
Set Membership: |
|

akt-1
Genomic Map